3-Isopropoxyphenylboronic acid 97%
Supplier: Apollo Scientific
Warning
OR3942-1G OR3942-25G OR3942-5G
APOSOR3942-1GEA 24.4 EUR
APOSOR3942-1G APOSOR3942-25G APOSOR3942-5G
3-Isopropoxyphenylboronic acid 97%
3-Isopropoxyphenylboronic acid
Formula: C₉H₁₃BO₃
Boiling Pt: 199 °C
Melting Pt: 76…81 °C
Density: 1.075 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03701512
CAS Number: 216485-86-8