4-Mercaptobenzonitrile

Supplier: Apollo Scientific
Warning

OR40005-1G OR40005-250MG
APOSOR40005-1GEA 190 EUR
APOSOR40005-1G APOSOR40005-250MG
4-Mercaptobenzonitrile
4-Mercaptobenzonitrile
Formula: C₇H₅NS
Boiling Pt: 118…119 °C/3mmHg
Melting Pt: 48…51 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00272354
CAS Number: 36801-01-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR