6-Iodoanthranilic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Amino-6-iodobenzoic acid

OR400135-100MG OR400135-1G OR400135-250MG
APOSOR400135-100MGEA 21.9 EUR
APOSOR400135-100MG APOSOR400135-1G APOSOR400135-250MG
6-Iodoanthranilic acid 98%
6-Iodoanthranilic acid
Formula: C₇H₆INO₂
MW: 263.0300 g/mol
Boiling Pt: 372 °C/760mmHg
Melting Pt: 145…147 °C
Flash Pt: 178.9 °C
Storage Temperature: Ambient
MDL Number: MFCD07774160
CAS Number: 20776-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR