4-Bromophthalic acid 98%
Supplier: Apollo Scientific
|
Warning
|
OR40201-25G
OR40201-100G
APOSOR40201-25GEA
87
EUR
APOSOR40201-25G
APOSOR40201-100G
4-Bromophthalic acid 98%
4-Bromophthalic acid
Formula:
C₈H₅BrO₄ Melting Pt: 162…164 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043538 CAS Number: 6968-28-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...