(R)-(-)-tert-Leucinol 98%
Supplier: Apollo Scientific
Warning
OR40208-1G OR40208-5G
APOSOR40208-1GEA 27.7 EUR
APOSOR40208-1G APOSOR40208-5G
(R)-(-)-tert-Leucinol 98%
(R)-(-)-tert-Leucinol
Formula: C₆H₁₅NO
Boiling Pt: 70 °C/0.4mmHg
Melting Pt: 30…33 °C
Density: 0.9 g/cm³ (25 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD01076574
CAS Number: 112245-09-7