Anthranilamide

Supplier: Apollo Scientific
Warning

Synonyms: 2-Aminobenzamide

OR4043-100G OR4043-10G OR4043-25G
APOSOR4043-100GEA 31.9 EUR
APOSOR4043-100G APOSOR4043-10G APOSOR4043-25G
Anthranilamide
Anthranilamide
Formula: C₇H₈N₂O
MW: 136.15 g/mol
Boiling Pt: 300 °C
Melting Pt: 110…114 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00007981
CAS Number: 88-68-6
EINECS: 201-851-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR