2,7-Dimethylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid
Supplier: Apollo Scientific
OR4078-1G OR4078-250MG
APOSOR4078-1GEA 85.7 EUR
APOSOR4078-1G APOSOR4078-250MG
2,7-Dimethylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid
2,7-Dimethylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid
Formula: C₉H₉N₃O₂
Melting Pt: 245…247 °C
Storage Temperature: Ambient
MDL Number: MFCD00067901
CAS Number: 175201-51-1