5-Bromo-1,3-benzodioxole 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromo-1,2-(methylenedioxy)benzene, 5-Bromobenzo[d][1,3]dioxole
OR4200-100G OR4200-25G OR4200-500G
APOSOR4200-100GEA 56.7 EUR
APOSOR4200-100G APOSOR4200-25G APOSOR4200-500G
5-Bromo-1,3-benzodioxole 98%
5-Bromo-1,3-benzodioxole
Formula: C₇H₅BrO₂
Boiling Pt: 85…86 °C/1mmHg
Density: 1.669 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00005821
CAS Number: 2635-13-4
EINECS: 220-123-5