2,6-Dichloro-3-nitropyridin-4-amine 97%
Supplier: Apollo Scientific
Warning
OR42145-1G OR42145-5G
APOSOR42145-1GEA 24.4 EUR
APOSOR42145-1G APOSOR42145-5G
2,6-Dichloro-3-nitropyridin-4-amine 97%
2,6-Dichloro-3-nitropyridin-4-amine
Formula: C₅H₃Cl₂N₃O₂
Melting Pt: 137…140 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03265754
CAS Number: 2897-43-0