2,4-Diiodo-1H-imidazole
Supplier: Apollo Scientific
Warning
OR42172-1G OR42172-2.5G OR42172-5G OR42172-10G
APOSOR42172-1GEA 221 EUR
APOSOR42172-1G APOSOR42172-2.5G APOSOR42172-5G APOSOR42172-10G
2,4-Diiodo-1H-imidazole
2,4-Diiodo-1H-imidazole
Formula: C₃H₂I₂N₂
Melting Pt: 184…186 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09800593
CAS Number: 19198-80-2