(S)-(-)-2,3-Epoxy-1-propanol 95%
Supplier: Apollo Scientific
Danger
Synonyms: (S)-(-)-Glycidol, (S)-Oxiran-2-ylmethanol
OR42180-100G OR42180-25G
APOSOR42180-100GEA 232 EUR
APOSOR42180-100G APOSOR42180-25G
(S)-(-)-2,3-Epoxy-1-propanol 95%
(S)-(-)-2,3-Epoxy-1-propanol
Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 66…67 °C/19mmHg
Density: 1.116 g/cm³ (25 °C)
Storage Temperature: Freezer
MDL Number: MFCD00074874
CAS Number: 60456-23-7
UN: 2810
ADR: 6.1 III