3,5-Dibromo-2-methylpyridine 97%
Supplier: Apollo Scientific
Warning
OR42204-1G OR42204-5G
APOSOR42204-1GEA 21.9 EUR
APOSOR42204-1G APOSOR42204-5G
3,5-Dibromo-2-methylpyridine 97%
3,5-Dibromo-2-methylpyridine
Formula: C6H5Br2N
MW: 250.9186 g/mol
Melting Pt: 22…23 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00234159
CAS Number: 38749-87-0