2-Iodo-2-methylpropane 95%
Supplier: Apollo Scientific
Danger
Synonyms: tert-Butyl iodide, 2-Methyl-2-propyliodide
OR42211-25G OR42211-100G
APOSOR42211-25GEA 143 EUR
APOSOR42211-25G APOSOR42211-100G
2-Iodo-2-methylpropane 95%
2-Iodo-2-methylpropane
Formula: C₄H₉I
MW: 184.02 g/mol
Boiling Pt: 99…100 °C
Melting Pt: –38 °C
Density: 1.547 g/cm³ (25 °C)
Flash Pt: 8 °C
Storage Temperature: Ambient
MDL Number: MFCD00001068
CAS Number: 558-17-8
EINECS: 209-190-1
UN: 2391
ADR: 3 II