2-Iodobenzonitrile 98%
Supplier: Apollo Scientific
Warning
OR4224-5G OR4224-25G OR4224-1G
APOSOR4224-5GEA 56.7 EUR
APOSOR4224-5G APOSOR4224-25G APOSOR4224-1G
2-Iodobenzonitrile 98%
2-Iodobenzonitrile
Formula: C₇H₄IN
Melting Pt: 51…55 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00079761
CAS Number: 4387-36-4
UN: 3439
ADR: 6.1,III