Tetrakis(triphenylphosphine)palladium(0)
Supplier: Apollo Scientific
Warning
Synonyms: Tetrakis(triphenylphosphine)palladium, Pd(PPh3)4
OR4225-1G OR4225-25G OR4225-5G
APOSOR4225-1GEA 27.7 EUR
APOSOR4225-1G APOSOR4225-25G APOSOR4225-5G
Tetrakis(triphenylphosphine)palladium(0)
Tetrakis(triphenylphosphine)palladium (0)
Formula: C₇₂H₆₀P₄Pd
MW: 1155.58 g/mol
Melting Pt: 103…107 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010012
CAS Number: 14221-01-3
EINECS: 238-086-9