2,5-Dichloropyridine
Supplier: Apollo Scientific
Warning
OR4274-25G OR4274-5G
APOSOR4274-25GEA 24.8 EUR
APOSOR4274-25G APOSOR4274-5G
2,5-Dichloropyridine
2,5-Dichloropyridine
Formula: C₅H₃Cl₂N
MW: 147.99 g/mol
Boiling Pt: 190…191 °C
Melting Pt: 59…62 °C
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00006239
CAS Number: 16110-09-1
EINECS: 240-278-2