2'-Bromo-4'-methylacetanilide 98%
Supplier: Apollo Scientific
Warning
OR4326-25G OR4326-5G OR4326-100G
APOSOR4326-25GEA 55.1 EUR
APOSOR4326-25G APOSOR4326-5G APOSOR4326-100G
2'-Bromo-4'-methylacetanilide 98%
2'-Bromo-4'-methylacetanilide
Formula: C₉H₁₀BrNO
Melting Pt: 118…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00017781
CAS Number: 614-83-5