(4-Cyanophenyl)methanesulphonyl chloride, Technical Grade
Supplier: Apollo Scientific
Danger
OR43515-1G OR43515-250MG
APOSOR43515-1GEA 125 EUR
APOSOR43515-1G APOSOR43515-250MG
(4-Cyanophenyl)methanesulphonyl chloride, Technical Grade
(4-Cyanophenyl)methanesulphonyl chloride
Formula: C₈H₆ClNO₂S
Melting Pt: 99.5…101.5 °C
Storage Temperature: Ambient
MDL Number: MFCD07690517
CAS Number: 56105-99-8
UN: 2923
ADR: 8+6.1,III