α,2-Dibromotoluene 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Bromobenzyl bromide
OR4356-1KG OR4356-250G OR4356-50G
APOSOR4356-1KGEA 348 EUR
APOSOR4356-1KG APOSOR4356-250G APOSOR4356-50G
α,2-Dibromotoluene 98%
α,2-Dibromotoluene
Formula: C₇H₆Br₂
MW: 249.93 g/mol
Boiling Pt: 129 °C/19mmHg
Melting Pt: 29…32 °C
Storage Temperature: Ambient
MDL Number: MFCD00000173
CAS Number: 3433-80-5
EINECS: 222-334-8
UN: 3261
ADR: 8 III