4-Bromophthalic anhydride 98%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Bromoisobenzofuran-1,3-dione
OR45117-100G OR45117-25G OR45117-500G
APOSOR45117-100GEA 48 EUR
APOSOR45117-100G APOSOR45117-25G APOSOR45117-500G
4-Bromophthalic anhydride 98%
4-Bromophthalic anhydride
Formula: C₈H₃BrO₃
MW: 227.01 g/mol
Boiling Pt: 301 °C
Melting Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00191323
CAS Number: 86-90-8