1-Methyl-1,4-cyclohexadiene, Technical Grade
Supplier: Apollo Scientific
Danger
OR451306-25G OR451306-5G OR451306-100G
APOSOR451306-25GEA 125 EUR
APOSOR451306-25G APOSOR451306-5G APOSOR451306-100G
1-Methyl-1,4-cyclohexadiene, Technical Grade
1-Methyl-1,4-cyclohexadiene
Formula: C₇H₁₀
Boiling Pt: 114…115 °C
Density: 0.838 g/cm³ (25 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00001538
CAS Number: 4313-57-9
UN: 3295
ADR: 3,II