2-Phenylethanethiol 95%
Supplier: Apollo Scientific
Warning
Synonyms: Phenethyl mercaptan
OR451326-100G OR451326-250G OR451326-25G OR451326-500G
APOSOR451326-100GEA 192 EUR
APOSOR451326-100G APOSOR451326-250G APOSOR451326-25G APOSOR451326-500G
2-Phenylethanethiol 95%
2-Phenylethanethiol
Formula: C₈H₁₀S
Boiling Pt: 217…218 °C
Density: 1.032 g/cm³ (25 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00004891
CAS Number: 4410-99-5