N-(tert-Butoxycarbonyl)-D-leucine hydrate 97%

Supplier: Apollo Scientific

OR452082-1G OR452082-25G OR452082-5G
APOSOR452082-1GEA 0 EUR
APOSOR452082-1G APOSOR452082-25G APOSOR452082-5G
N-(tert-Butoxycarbonyl)-D-leucine hydrate 97%
N-(tert-Butoxycarbonyl)-D-leucine hydrate
Formula: C₁₁H₂₁NO₄
Storage Temperature: Ambient
MDL Number: MFCD00038294
CAS Number: 16937-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR