Ethyl-3-iodobenzoate
Supplier: Apollo Scientific
Warning
Synonyms: 3-Iodobenzoic acid ethyl ester
OR4534-100G OR4534-500G
APOSOR4534-100GEA 79.9 EUR
APOSOR4534-100G APOSOR4534-500G
Ethyl-3-iodobenzoate
Ethyl 3-iodobenzoate
Formula: C₉H₉IO₂
Boiling Pt: 160…161 °C/38mmHg
Density: 1.66 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00044769
CAS Number: 58313-23-8