1-Iodopropane
Supplier: Apollo Scientific
Warning
Synonyms: 1-Propyl iodide
OR4541-25G OR4541-5G OR4541-100G
APOSOR4541-25GEA 36.4 EUR
APOSOR4541-25G APOSOR4541-5G APOSOR4541-100G
1-Iodopropane
1-Iodopropane
Formula: C₃H₇I
MW: 169.99 g/mol
Boiling Pt: 101…102 °C
Melting Pt: –101 °C
Density: 1.743 g/cm³ (25 °C)
Flash Pt: 44 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001095
CAS Number: 107-08-4
UN: 2392
ADR: 3 III
Merck Index: 12,08047