ω-Bromo-2-methoxyacetophenone 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Methoxyphenacyl bromide, α-Bromo-2-methoxyacetophenone, 2-Bromo-2'-methoxyacetophenone, 2-Bromo-1-(2-methoxyphenyl)ethanone, 2-Bromo-1-(2-methoxyphenyl)ethan-1-one, Bromomethyl o-methoxyphenyl ketone
OR4557-100G OR4557-25G OR4557-5G
APOSOR4557-100GEA 389 EUR
APOSOR4557-100G APOSOR4557-25G APOSOR4557-5G
ω-Bromo-2-methoxyacetophenone 97%
ω-Bromo-2-methoxyacetophenone
Formula: C₉H₉BrO₂
Melting Pt: 43…45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000196
CAS Number: 31949-21-0
EINECS: 250-870-2
UN: 3261
ADR: 8,II