2-Bromo-5-iodotoluene
Supplier: Apollo Scientific
Warning
OR4591-25G OR4591-5G OR4591-100G
APOSOR4591-25GEA 29 EUR
APOSOR4591-25G APOSOR4591-5G APOSOR4591-100G
2-Bromo-5-iodotoluene
2-Bromo-5-iodotoluene
Formula: C₇H₆BrI
Boiling Pt: 148 °C/20mmHg
Density: 2.07 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00070749
CAS Number: 202865-85-8