5-Bromo-2-iodotoluene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-4-iodo-3-methylbenzene
OR4595-100G OR4595-250G
APOSOR4595-100GEA 74.4 EUR
APOSOR4595-100G APOSOR4595-250G
5-Bromo-2-iodotoluene 98%
5-Bromo-2-iodotoluene
Formula: C₇H₆BrI
MW: 296.93 g/mol
Boiling Pt: 262…265 °C
Density: 2.08 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00060664
CAS Number: 116632-39-4