5-Bromo-2-iodo-m-xylene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Bromo-1,3-dimethyl-2-iodobenzene
OR4596-100G OR4596-25G OR4596-5G
APOSOR4596-100GEA 154 EUR
APOSOR4596-100G APOSOR4596-25G APOSOR4596-5G
5-Bromo-2-iodo-m-xylene 98%
5-Bromo-2-iodo-m-xylene
Formula: C₈H₈BrI
Melting Pt: 42…44 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094656
CAS Number: 206559-43-5