3-Iodopropene
Supplier: Apollo Scientific
Danger
Synonyms: 3-Iodo-1-Propene, Allyl iodide, 3-Propenyl iodide
OR460034-25G OR460034-5G
APOSOR460034-25GEA 118 EUR
APOSOR460034-25G APOSOR460034-5G
3-Iodopropene
3-Iodopropene
Formula: C₃H₅I
MW: 167.98 g/mol
Boiling Pt: 102…103 °C
Melting Pt: –99 °C
Density: 1.832 g/cm³ (25 °C)
Flash Pt: 18 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001094
CAS Number: 556-56-9
EINECS: 209-130-4
UN: 1723
ADR: 3+8 II
Merck Index: 13,00291