4,4,5,5-Tetrachloro-1,3-dioxolan-2-one
Supplier: Apollo Scientific
OR460071-1G OR460071-5G
APOSOR460071-1GEA 310 EUR
APOSOR460071-1G APOSOR460071-5G
4,4,5,5-Tetrachloro-1,3-dioxolan-2-one
4,4,5,5-Tetrachloro-1,3-dioxolan-2-one
Formula: C₃Cl₄O₃
Storage Temperature: Ambient
MDL Number: MFCD00030631
CAS Number: 22432-68-4