1-(Methylsulphonyl)piperidine-4-carbaldehyde
Supplier: Apollo Scientific
Warning
OR46015-1G OR46015-250MG OR46015-100MG OR46015-500MG OR46015-2.5G OR46015-5G
APOSOR46015-1GEA 519 EUR
APOSOR46015-1G APOSOR46015-250MG APOSOR46015-100MG APOSOR46015-500MG APOSOR46015-2.5G APOSOR46015-5G
1-(Methylsulphonyl)piperidine-4-carbaldehyde
1-(Methylsulphonyl)piperidine-4-carbaldehyde
Formula: C₇H₁₃NO₃S
MW: 191.25 g/mol
Melting Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD11582937
CAS Number: 241134-35-0