2-Tolualdehyde 98%
Supplier: Apollo Scientific
Danger
Synonyms: o-Tolualdehyde, 2-Methylbenzaldehyde
OR46026-100G OR46026-1KG OR46026-500G OR46026-25G
APOSOR46026-100GEA 43.5 EUR
APOSOR46026-100G APOSOR46026-1KG APOSOR46026-500G APOSOR46026-25G
2-Tolualdehyde 98%
2-Tolualdehyde
Formula: C₈H₈O
MW: 120.15 g/mol
Boiling Pt: 199…200 °C
Melting Pt: –35 °C
Density: 1.037 g/cm³ (25 °C)
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00003338
CAS Number: 529-20-4
EINECS: 208-452-2
Merck Index: 12,09665