cis-1,3-Cyclopentanedicarboxylic acid
Supplier: Apollo Scientific
Warning
Synonyms: cis-Cyclopentane-1,3-dicarboxylic acid
OR46133-1G OR46133-5G
APOSOR46133-1GEA 30.6 EUR
APOSOR46133-1G APOSOR46133-5G
cis-1,3-Cyclopentanedicarboxylic acid
cis-1,3-Cyclopentanedicarboxylic acid
Formula: C₇H₁₀O₄
MW: 158.16 g/mol
Melting Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD01311175
CAS Number: 876-05-1