1,2,4-Trimethoxybenzene 98%
Supplier: Apollo Scientific
Warning
OR4622-100G OR4622-10G OR4622-50G
APOSOR4622-100GEA 63.8 EUR
APOSOR4622-100G APOSOR4622-10G APOSOR4622-50G
1,2,4-Trimethoxybenzene 98%
1,2,4-Trimethoxybenzene
Formula: C₉H₁₂O₃
Boiling Pt: 247 °C
Density: 1.126 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008360
CAS Number: 135-77-3
EINECS: 205-219-7