Furan-2,5-diyldimethanol 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2,5-Furandimethanol, 5-(Hydroxymethyl)furfuryl Alcohol
OR46504-10G OR46504-5G OR46504-25G OR46504-100G OR46504-500G
APOSOR46504-10GEA 89.9 EUR
APOSOR46504-10G APOSOR46504-5G APOSOR46504-25G APOSOR46504-100G APOSOR46504-500G
Furan-2,5-diyldimethanol 97%
Furan-2,5-diyldimethanol
Formula: C₆H₈O₃
Boiling Pt: 95…120 °C/0.4mmHg
Melting Pt: 74…77 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003253
CAS Number: 1883-75-6