4,6-Dimethyl-2-(1H-pyrrol-1-yl)pyrimidine
Supplier: Apollo Scientific
OR46714-1G OR46714-250MG OR46714-500MG OR46714-5G
APOSOR46714-1GEA 328 EUR
APOSOR46714-1G APOSOR46714-250MG APOSOR46714-500MG APOSOR46714-5G
4,6-Dimethyl-2-(1H-pyrrol-1-yl)pyrimidine
4,6-Dimethyl-2-(1H-pyrrol-1-yl)pyrimidine
Formula: C₁₀H₁₁N₃
Storage Temperature: Ambient
MDL Number: MFCD02665508
CAS Number: 60795-33-7