5-Bromoisophthalaldehyde 98%

Supplier: Apollo Scientific
Warning

OR46757-1G OR46757-25G OR46757-5G
APOSOR46757-1GEA 34.8 EUR
APOSOR46757-1G APOSOR46757-25G APOSOR46757-5G
5-Bromoisophthalaldehyde 98%
5-Bromoisophthalaldehyde
Formula: C₈H₅BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD08690086
CAS Number: 120173-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR