1-Iodododecane

Supplier: Apollo Scientific
Warning

Synonyms: 1-Dodecyl iodide

OR4679-1G OR4679-5G
APOSOR4679-1GEA 14.7 EUR
APOSOR4679-1G APOSOR4679-5G
1-Iodododecane
1-Iodododecane
Formula: C₁₂H₂₅I
MW: 296.24 g/mol
Boiling Pt: 159…160 °C/15mmHg
Melting Pt: –3 °C
Density: 1.201 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001088
CAS Number: 4292-19-7
EINECS: 224-293-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR