2-(2-Chloroethoxy)-4'-nitroacetanilide
Supplier: Apollo Scientific
OR470179-1G OR470179-5G
APOSOR470179-1GEA 286 EUR
APOSOR470179-1G APOSOR470179-5G
2-(2-Chloroethoxy)-4'-nitroacetanilide
2-(2-Chloroethoxy)-4'-nitroacetanilide
Formula: C₁₀H₁₁ClN₂O₄
Storage Temperature: Ambient
MDL Number: MFCD18379274
CAS Number: 811450-82-5