4-Cyanobenzenecarboximidamide hydrochloride
Supplier: Apollo Scientific
Synonyms: 4-Cyanobenzamidine hydrochloride
OR471156-1G OR471156-250MG
APOSOR471156-1GEA 804 EUR
APOSOR471156-1G APOSOR471156-250MG
4-Cyanobenzenecarboximidamide hydrochloride
4-Cyanobenzenecarboximidamide hydrochloride
Formula: C₈H₈ClN₃
Storage Temperature: Ambient
MDL Number: MFCD16622253
CAS Number: 117837-77-1