2-(1-Piperidinyl)acetamidoxime 97%
Supplier: Apollo Scientific
Warning
OR4739-1G OR4739-5G OR4739-250MG
APOSOR4739-1GEA 116 EUR
APOSOR4739-1G APOSOR4739-5G APOSOR4739-250MG
2-(1-Piperidinyl)acetamidoxime 97%
2-(1-Piperidinyl)acetamidoxime
Formula: C₇H₁₅N₃O
Melting Pt: 154 °C
Storage Temperature: Ambient
MDL Number: MFCD00052662
CAS Number: 175136-64-8