5-Vinyluracil 95%
Supplier: Apollo Scientific
Synonyms:
5-Vinylpyrimidine-2,4(1H,3H)-dione
OR480214-1G
OR480214-500MG
OR480214-5G
APOSOR480214-1GEA
855
EUR
APOSOR480214-1G
APOSOR480214-500MG
APOSOR480214-5G
5-Vinyluracil 95%
5-Vinyluracil
Formula:
C₆H₆N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00913267 CAS Number: 37107-81-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...