5-Chloro-2-furaldehyde 98%
Supplier: Apollo Scientific
OR480749-25G
OR480749-5G
APOSOR480749-25GEA
127
EUR
APOSOR480749-25G
APOSOR480749-5G
5-Chloro-2-furaldehyde 98%
5-Chloro-2-furaldehyde
Formula:
C₅H₃ClO₂ Storage Temperature: Ambient |
MDL Number:
MFCD02752599 CAS Number: 21508-19-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...