2-(Trimethylsilyl)ethanol
Supplier: Apollo Scientific
Warning
OR4827-100G OR4827-25G OR4827-5G
APOSOR4827-100GEA 120 EUR
APOSOR4827-100G APOSOR4827-25G APOSOR4827-5G
2-(Trimethylsilyl)ethanol
2-(Trimethylsilyl)ethanol
Formula: HOH₂CCH₂Si(CH₃)₃
Boiling Pt: 71…73 °C
Density: 0.825 g/cm³ (25 °C)
Flash Pt: 56 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002825
CAS Number: 2916-68-9
EINECS: 220-844-5
UN: 1987
ADR: 3,III