1,2-Epoxycyclopentane
Supplier: Apollo Scientific
Danger
OR4849-25G OR4849-5G
APOSOR4849-25GEA 0 EUR
APOSOR4849-25G APOSOR4849-5G
1,2-Epoxycyclopentane
1,2-Epoxycyclopentane
Formula: C₅H₈O
Boiling Pt: 101…102 °C
Density: 0.965 g/cm³ (25 °C)
Flash Pt: 10 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005161
CAS Number: 285-67-6
EINECS: 206-005-6
UN: 1993
ADR: 3,II