Pyromellitic dianhydride 98%
Supplier: Apollo Scientific
Danger
Synonyms: 1,2,4,5-Benzenetetracarboxylic anhydride
OR4903-100G OR4903-2.5KG OR4903-500G
APOSOR4903-100GEA 21.9 EUR
APOSOR4903-100G APOSOR4903-2.5KG APOSOR4903-500G
Pyromellitic dianhydride 98%
Pyromellitic dianhydride
Formula: C₁₀H₂O₆
Boiling Pt: 397…400 °C
Melting Pt: 283…288 °C
Density: 1.68 g/cm³ (25 °C)
Flash Pt: 380 °C
Storage Temperature: Ambient
MDL Number: MFCD00005005
CAS Number: 89-32-7
EINECS: 201-898-9