2,2-Dimethylpropane-1,3-diamine
Supplier: Apollo Scientific
Danger
Synonyms: 2,2-Dimethyl-1,3-propanediamine
OR4925-100G OR4925-25G
APOSOR4925-100GEA 116 EUR
APOSOR4925-100G APOSOR4925-25G
2,2-Dimethylpropane-1,3-diamine
2,2-Dimethylpropane-1,3-diamine
Formula: C₅H₁₄N₂
MW: 102.1800 g/mol
Boiling Pt: 152…154 °C
Melting Pt: 29…31 °C
Density: 0.851 g/cm³ (25 °C)
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00009801
CAS Number: 7328-91-8
EINECS: 230-819-0
UN: 2921
ADR: 8+4.1 II