3-Bromotoluene 99%
Supplier: Apollo Scientific
Warning
OR4938-100G OR4938-25G OR4938-500G
APOSOR4938-100GEA 75.4 EUR
APOSOR4938-100G APOSOR4938-25G APOSOR4938-500G
3-Bromotoluene 99%
3-Bromotoluene
Formula: C₇H₇Br
Boiling Pt: 182…184 °C
Melting Pt: –40 °C
Density: 1.41 g/cm³ (25 °C)
Flash Pt: 59 °C
Storage Temperature: Ambient
MDL Number: MFCD00000085
CAS Number: 591-17-3
EINECS: 209-702-3
UN: 1993
ADR: 3,III