4-Bromobenzoic acid 98%
Supplier: Apollo Scientific
Warning
OR4997-100G OR4997-25G OR4997-500G
APOSOR4997-100GEA 24.8 EUR
APOSOR4997-100G APOSOR4997-25G APOSOR4997-500G
4-Bromobenzoic acid 98%
4-Bromobenzoic acid
Formula: C₇H₅BrO₂
Melting Pt: 252…258 °C
Density: 1.894 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002529
CAS Number: 586-76-5
EINECS: 209-581-7